Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Amino-3,3-dimethyl-1,1-diphenyl-1-butanol 98.0+%, TCI America™
SDP

Catalog No. p-7117947 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Catalog No. A25031G

Supplier: TCI America A25031G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 144054-70-6
Molecular Formula C18H23NO
Molecular Weight (g/mol) 269.388
MDL Number MFCD03427198
InChI Key HZIHDWNOPKIOCK-INIZCTEOSA-N
PubChem CID 10333308
IUPAC Name (2S)-2-amino-3,3-dimethyl-1,1-diphenylbutan-1-ol
SMILES CC(C)(C)C(C(C1=CC=CC=C1)(C2=CC=CC=C2)O)N

Specifications

Melting Point 134°C
Color White
Quantity 1 g
Formula Weight 269.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-2-Amino-3,3-dimethyl-1,1-diphenyl-1-butanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.