Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Azetidinecarboxylic acid, 99+%

Catalog No. p-3790836
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105141000 100 mg Glass bottle
AC105142500 250 mg Glass bottle
AC105140010 1 g Glass bottle
AC105140050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC105141000 Supplier Thermo Scientific Chemicals Supplier No. 105141000
Only null left
Add to Cart

CAS: 2133-34-8 | C4H7NO2 | 101.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2133-34-8
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.1
InChI Key IADUEWIQBXOCDZ-VKHMYHEASA-N
Synonym l-azetidine-2-carboxylic acid, s-azetidine-2-carboxylic acid, s---2-azetidinecarboxylic acid, 2s-azetidine-2-carboxylic acid, azetidyl-2-carboxylic acid, l-azetidine 2-carboxylic acid, s-2-azetidinecarboxylic acid, 2-azetidinecarboxylic acid, s, s---azetidine-2-carboxylic acid, 2-azetidinecarboxylic acid, l
PubChem CID 16486
ChEBI CHEBI:6198
IUPAC Name (2S)-azetidine-2-carboxylic acid
SMILES C1CNC1C(=O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 206°C to 207°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 100 mg
Merck Index 10,911
Specific Rotation Condition −125° (25°C c=1,water)
Specific Rotation -125
Solubility Information Solubility in water: 5% in water
Formula Weight 101.1
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (S)-(-)-2-Azetidinecarboxylic acid
Show More Show Less
EINECSNumber 218-362-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.