Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Methylbutylamine, 98+%

Catalog No. AAL14063MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14063MD 250 mg
AAL1406303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14063MD Supplier Thermo Scientific Chemicals Supplier No. L14063MD

CAS: 34985-37-0 | C5H13N | 87.166 g/mol

(S)-(-)-2-Methylbutylamine is used as aintermediate in organic synthesis, as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-2-Methylbutylamine is used as aintermediate in organic synthesis, as pharmaceutical intermediate.

Solubility
Fully miscible in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, carbondioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34985-37-0
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00064430
InChI Key VJROPLWGFCORRM-YFKPBYRVSA-N
Synonym s---2-methylbutylamine, s-2-methylbutylamine, unii-d8q8rzf45x, 2s-2-methylbutan-1-amine, s---2-methyl-1-butylamine, d8q8rzf45x, 2s-2-methyl-butan-1-amine, 2-methyl-1-butanamine #, s---1-amino-2-methylbutane hydrochloride
PubChem CID 2724272
IUPAC Name (2S)-2-methylbutan-1-amine
SMILES CCC(C)CN

Specifications

Boiling Point 96°C to 97°C
Flash Point 3°C (37°F)
Quantity 250 mg
UN Number UN2733
Beilstein 1718807
Optical Rotation −5.9° (Neat)
Formula Weight 87.16
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-2-Methylbutylamine
Hazard Category Danger
Hazard Statement GHS H Statement
H225-H314-H318-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.