Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(S)-(-)-2-Phenyl-1-propanol, 99+%, Thermo Scientific™
Click to view available options
Quantity:
1 mL
Packaging:
Glass bottle
Specifications
Specifications
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Color | Colorless |
Boiling Point | 214°C |
Flash Point | 108°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.5252 to 1.5272 |
Linear Formula | C6H5CH(CH3)CH2OH |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Harmful if swallowed. |
Precautionary Statement | GHS P Statement IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction