Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ S 24795
GREENER_CHOICE

Catalog No. 351810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-181-0 10 mg
35-185-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-181-0 Supplier Tocris Bioscience™ Supplier No. 3518/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Partial agonist at α7 nAChR

Partial agonist of α7 nAChR. Improves mnemonic function in aged mice for the treatment of aging-related memory disturbances. Displays more efficient memory-enhancing effects than memantine (Cat. No. 0773) in Alzheimer's disease mouse models.

Chemical Identifiers

CAS 304679-75-2
Molecular Formula C14H13BrINO

Specifications

Quantity 10 mg
Formula Weight 418.07
Purity >98%
Molecular Weight (g/mol) 418.072
Chemical Name or Material S 24795
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.