Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-Amino-5-hexynoic acid hydrochloride, 95%

Catalog No. p-7034329
Click to view available options
Quantity:
250 mg
1 g

CAS: 270596-46-8 | C6H10ClNO2 | 163.601 g/mol

(S)-3-Amino-5-hexynoic acid hydrochloride is used for labeling and detection purpose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-Amino-5-hexynoic acid hydrochloride is used for labeling and detection purpose.

Solubility
Soluble in water (90 g/L at 25°C).

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 270596-46-8
Molecular Formula C6H10ClNO2
Molecular Weight (g/mol) 163.601
MDL Number MFCD01861006
InChI Key FVKOZZHCHSRKJA-JEDNCBNOSA-N
Synonym s-3-amino-5-hexynoic acid hydrochloride, s-3-aminohex-5-ynoic acid hydrochloride, 3s-3-aminohex-5-ynoic acid hydrochloride, s-3-amino-5-hexynoic acid-hcl, l-beta-homopropargylglycine hydrochloride, 5-hexynoic acid, 3-amino-, 3s-, hydrochloride 1:1, s-3-amino-5-hexynoic acidhcl, l-ss-homopropargylglycine hydrochloride
PubChem CID 2761700
IUPAC Name (3S)-3-aminohex-5-ynoic acid;hydrochloride
SMILES C#CCC(CC(=O)O)N.Cl

Specifications

Solubility Information Soluble in water (90g/L at 25°C).
Quantity 250 mg
Formula Weight 163.6
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material (S)-3-Amino-5-hexynoic acid hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.