Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-(Methylamino)-1-(2-thienyl)-1-propanol 98.0+%, TCI America™
SDP

Catalog No. M215725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M215725G 25 g
M21575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M215725G Supplier TCI America Supplier No. M215725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 116539-55-0
Molecular Formula C8H13NOS
Molecular Weight (g/mol) 171.258
MDL Number MFCD07357308
InChI Key YEJVVFOJMOHFRL-ZETCQYMHSA-N
Synonym (S)-2-[3-(Methylamino)-1-hydroxypropyl]thiophene
PubChem CID 10095047
IUPAC Name (1S)-3-(methylamino)-1-thiophen-2-ylpropan-1-ol
SMILES CNCCC(C1=CC=CS1)O

Specifications

Melting Point 74°C
Quantity 25 g
Formula Weight 171.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-3-(Methylamino)-1-(2-thienyl)-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.