Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-4-Penten-2-ol, 97%

Catalog No. p-7028417
Click to view available options
Quantity:
1 g
5 g

CAS: 55563-79-6 | C5H10O | 86.134 g/mol

(S)-(+)-4-Penten-2-ol is involved in the synthesis of goniothalamin, hexadecanolide, massoia lactone, and parasorbic acid through sequential allylboration-esterification ring-closing metathesis reactions. It is also used to prepare 2-pentanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-4-Penten-2-ol is involved in the synthesis of goniothalamin, hexadecanolide, massoia lactone, and parasorbic acid through sequential allylboration-esterification ring-closing metathesis reactions. It is also used to prepare 2-pentanone.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55563-79-6
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD03427200
InChI Key ZHZCYWWNFQUZOR-YFKPBYRVSA-N
Synonym s-+-4-penten-2-ol, 4-penten-2-ol, 2s, 2s-pent-4-en-2-ol, 2s-4penten-2-ol, s-4-pentene-2-ol, 2s-4-penten-2-ol, s-pent-4-en-2-ol, s-2-hydroxypent-4-ene
PubChem CID 6994332
IUPAC Name (2S)-pent-4-en-2-ol
SMILES CC(CC=C)O

Specifications

Density 0.837
Boiling Point 115°C to 116°C
Flash Point 30°C (86°F)
Refractive Index 1.425
Quantity 1 g
UN Number UN1987
Optical Rotation +5.2° (c=1 in Chloroform)
Formula Weight 86.13
Percent Purity 97%
Chemical Name or Material (S)-(+)-4-Penten-2-ol
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.