Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S-Adenosyl-Homocysteine Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA185037

Catalog No. PA185037


Only null left
Add to Cart

Description

Description

Rabbit anti S-adenosyl homosysteine antibody recognizes S-Adenosyl Homocysteine, an intermediate in the synthesis of the amino acid cysteine.

S-Adenosylhomocysteine (AdoHcy) is the immediate precursor of all of the homocysteine produced in the body. The reaction is catalyzed by S-adenosylhomocysteine hydrolase and is reversible with the equilibrium favoring formation of AdoHcy. In vivo, the reaction is driven in the direction of homocysteine formation by the action of the enzyme adenosine deaminase, which converts the second product of the S-adenosylhomocysteine hydrolase reaction, adenosine, to inosine. Except for methyl transfer from betaine and from methylcobalamin in the methionine synthase reaction, AdoHcy is the product of all methylation reactions that involve S-adenosylmethionine (AdoMet) as the methyl donor. Methylation is significant in epigenetic regulation of protein expression via DNA and histone methylation. The inhibition of these AdoMet-mediated processes by AdoHcy is a proven mechanism for metabolic alteration. Because the conversion of AdoHcy to homocysteine is reversible, with the equilibrium favoring the formation of AdoHcy, increases in plasma homocysteine are accompanied by an elevation of AdoHcy in most cases. Disturbances in the transmethylation pathway indicated by abnormal S-adenosylmethionine, S-adenosylhomocysteine or their ratio have been reported in many neurodegenerative diseases, such as dementia, depression or Parkinson's disease. (PMID: 18065573, 17892439). [National Center for Biotechnology Information. PubChem Compound Database; CID=439155].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

S-Adenosyl-Homocysteine
Polyclonal
Unconjugated
AdoHcy; C14H20N6O5S; Formycinylhomocysteine; S-(5'-adenosyl)-L-homocysteine; S-adenosylhomocysteine; S-adenosyl-L-homocysteine
S-Adenosyl-Homocysteine conjugated to KLH.
1 mL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Chemical
ELISA
5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Rabbit
Protein G
RUO
Chemical
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.