Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-alpha-Allylglycine, 98%, 98% ee

Catalog No. AC275410010
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC275410010 1 g Glass bottle
AC275412500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC275410010 Supplier Thermo Scientific Chemicals Supplier No. 275410010
Add to Cart

CAS: 16338-48-0 | C5H9NO2 | 115.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16338-48-0
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD00002627
InChI Key WNNNWFKQCKFSDK-BYPYZUCNSA-N
Synonym l-allylglycine, s-2-aminopent-4-enoic acid, 2s-2-aminopent-4-enoic acid, h-gly ally-oh, s---2-amino-4-pentenoic acid, s-2-amino-4-pentenoic acid, s-allylglycine, l-2-amino-4-pentenoic acid, 3-vinyl-l-alanine
PubChem CID 167529
IUPAC Name (2S)-2-aminopent-4-enoic acid
SMILES C=CCC(C(=O)O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 253.0°C to 255.0°C
Color White
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2C=CHCH2CH(NH2)CO2H
Quantity 1 g
Beilstein 04, 2852
Specific Rotation Condition −37° (20°C c=4,H2O)
Specific Rotation − 37.00
Solubility Information Solubility in water: soluble.
Formula Weight 115.13
Percent Purity 98%,98% ee
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-α-Allylglycine, 98%, (98% e.e.)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.