Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium carbonate decahydrate, 99+%, extra pure

Catalog No. AC271070025
Change view
Click to view available options
Quantity:
100 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC271070025 2.5 kg Plastic Bottle
AC271071000 100 g Plastic Bottle
AC271070100 10 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC271070025 Supplier Thermo Scientific Chemicals Supplier No. 271070025
Only null left
Add to Cart

CAS: 2-1-6132 | CNa2O3·10H2O | 286.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-1-6132
Molecular Formula CNa2O3·10H2O
Molecular Weight (g/mol) 286.14
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

Specifications

Melting Point 34.0°C
Color White
Physical Form Crystals
Quantity 2.5 kg
Assay Percent Range 99+%
Linear Formula Na2CO3·10H2O
Fieser 7322
Merck Index 158731
Solubility Information Solubility in water: 210g/L (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
Percent Purity ≥99%
Grade Extra Pure
Packaging Plastic Bottle
Specific Gravity 1.46
Density 1.46 g/mL
Chemical Name or Material Sodium carbonate decahydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.