Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ S-arrestin Monoclonal Antibody (PDS-1), HRP
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA516030
Description
This antibody reacts weakly with rat samples. Suggested positive control: retina lysate.
S-antigen, also identified as arrestin, is a protein involved in the regulation of phototransduction.
Specifications
S-arrestin | |
Monoclonal | |
0.68 mg/mL | |
PBS with no preservative | |
P08168, P10523, P79260 | |
SAG | |
Porcine retinal S-antigen (arrestin). | |
100 μL | |
Primary | |
Human, Bovine, Pig | |
Antibody | |
IgG1 |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
PDS-1 | |
HRP | |
SAG | |
48 kDa protein; A930001K18Rik; Arr1; arrestin; arrestin 1; arrestin1; arrestin-1; DKFZp686D1084; DKFZp686I1383; Irbp; photoreceptor protein/alleged cGMP phosphodiesterase activation modulator; put. S-antigen C-terminus; retinal S antigen; retinal S-antigen; retinal S-antigen (48 KDa protein); rod arrestin; rod photoreceptor arrestin; RP47; Sag; S-AG; SAGMR; SANTI; S-antigen; S-antigen visual arrestin; S-antigen, retina and pineal gland (arrestin); S-antigen; retina and pineal gland (arrestin); S-arrestin; S-arrestin short form; visual arrestin 1 | |
Mouse | |
Protein G | |
RUO | |
280922, 397151, 6295 | |
4°C, store in dark | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction