Learn More
Invitrogen™ S-arrestin Monoclonal Antibody (PDS1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA3814
Description
MA3-814 detects Visual Arrestin from human, rat, porcine, bovine, ovine, and goat tissues. MA3-814 has been successfully used in Immunohistochemistry, ELISA, and Western blotting procedures. By Western blot it detects a 37-kDa band representing Visual arrestin. The MA3-814 antigen is Visual arrestin isolated from Porcine cells.
S-antigen, also identified as arrestin, is a protein involved in the regulation of phototransduction.
Specifications
S-arrestin | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.05% sodium azide | |
P08168, P10523, P15887, P79260 | |
SAG | |
Visual Arrestin isolated from Porcine Cells. | |
RUO | |
101105037, 102189368, 25539, 280922, 397151, 6295 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
PDS1 | |
Unconjugated | |
SAG | |
48 kDa protein; A930001K18Rik; Arr1; arrestin; arrestin 1; arrestin1; arrestin-1; DKFZp686D1084; DKFZp686I1383; Irbp; photoreceptor protein/alleged cGMP phosphodiesterase activation modulator; put. S-antigen C-terminus; retinal S antigen; retinal S-antigen; retinal S-antigen (48 KDa protein); rod arrestin; rod photoreceptor arrestin; RP47; Sag; S-AG; SAGMR; SANTI; S-antigen; S-antigen visual arrestin; S-antigen, retina and pineal gland (arrestin); S-antigen; retina and pineal gland (arrestin); S-arrestin; S-arrestin short form; visual arrestin 1 | |
Mouse | |
100 μL | |
Primary | |
Human, Rat, Bovine, Sheep, Pig, Goat | |
Antibody | |
IgG1 |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.