Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-Benzylsuccinic Acid 98.0+%, TCI America™
SDP

Catalog No. B414225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B414225G 25 g
B41425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B414225G Supplier TCI America Supplier No. B414225G

Chemical Identifiers

CAS 3972-36-9
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00798341
InChI Key GTOFKXZQQDSVFH-VIFPVBQESA-N
PubChem CID 446168
IUPAC Name (2S)-2-benzylbutanedioic acid
SMILES C1=CC=C(C=C1)CC(CC(=O)O)C(=O)O

Specifications

Melting Point 166°C
Color White
Quantity 25 g
Formula Weight 208.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-Benzylsuccinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.