Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ (S)-(-)-Blebbistatin
GREENER_CHOICESDP

Catalog No. 18521 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-521 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-521 Supplier R&D Systems™ Supplier No. 1852/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of myosin II ATPase activity. Active enantiomer of (±)-blebbistatin (Cat. No. 1760)

(S)-(-)-Blebbistatin is a selective inhibitor of myosin II ATPase activity (IC50 ∼0.5 - 5 μM); active enantiomer of (±)-blebbistatin. Inhibits contraction of the cleavage furrow without disrupting mitosis or contractile ring assembly. (S)-(-)-Blebbistatin rapidly and reversibly blocks cell blebbing, and disrupts directed cell migration and cytokinesis in vertebrate cells.

Inactive Enantiomer and Racemate also available.

Chemical Identifiers

CAS 856925-71-8
Molecular Formula C18H16N2O2

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 292.34
Purity >99%
Molecular Weight (g/mol) 292.34
Chemical Name or Material (S)-(-)-Blebbistatin
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.