Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ S-Boc-2-mercapto-4,6-dimethylpyrimidine, 97%
Click to view available options
Quantity:
5g
25g
Description
Applications
S-Boc-2-mercapto-4,6-dimethylpyrimidine can react with propylamine to produce propyl-carbamic acid tert-butyl ester. It is used as a pharmaceutical intermediate.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
S-Boc-2-mercapto-4,6-dimethylpyrimidine can react with propylamine to produce propyl-carbamic acid tert-butyl ester. It is used as a pharmaceutical intermediate.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications
Quantity | 5g |
Melting Point | 48°C to 51°C |
Flash Point | >110°C (230°F) |
Beilstein | 396984 |
Formula Weight | 240.33 |
Percent Purity | 97% |
Chemical Name or Material | S-Boc-2-mercapto-4,6-dimethylpyrimidine |
Safety and Handling
EINECSNumber : 255-562-1
TSCA : No
Recommended Storage : Keep cold