Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Butyrylthiocholine iodide, 98%

Catalog No. p-7020608
Click to view available options
Quantity:
5 g
25 g

CAS: 1866-16-6 | C9H20INOS | 317.229 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1866-16-6
Molecular Formula C9H20INOS
Molecular Weight (g/mol) 317.229
MDL Number MFCD00011845
InChI Key WEQAAFZDJROSBF-UHFFFAOYSA-M
Synonym butyrylthiocholine iodide, s-butyrylthiocholine iodide, 2-butyrylthio-n,n,n-trimethylethanaminium iodide, s-n-butyrylthiocholine iodide, 2-mercaptoethyl trimethylammonium iodide butyrate, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide, propylcarbonylthioethyl trimethylammonium iodide, 1-2-trimethylamino ethylthio butan-1-one, iodide, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide 1:1
PubChem CID 74630
IUPAC Name 2-butanoylsulfanylethyl(trimethyl)azanium;iodide
SMILES CCCC(=O)SCC[N+](C)(C)C.[I-]

Specifications

Melting Point 172°C to 176°C
Linear Formula CH3CH2CH2COSCH2CH2N(CH3)3I
Quantity 5 g
Beilstein 3729509
Sensitivity Hygroscopic; Light sensitive
Formula Weight 317.23
Percent Purity 98%
Chemical Name or Material S-Butyrylthiocholine iodide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-475-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.