Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (S)-(-)-Carbidopa
GREENER_CHOICESDP

Catalog No. 045525 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-552-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 04-552-5 Supplier Tocris Bioscience™ Supplier No. 0455/25

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Aromatic L-amino acid decarboxylase inhibitor

Peripheral decarboxylase inhibitor, used in combination with levodopa for treatment of Parkinsonism.

Chemical Identifiers

CAS 28860-95-9
Molecular Formula C10H14N2O4

Specifications

Quantity 25 mg
Formula Weight 226.23
Purity >99%
Molecular Weight (g/mol) 226.232
Chemical Name or Material (S)-(-)-Carbidopa
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.