Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (S)-Crizotinib, >98%
GREENER_CHOICE

Catalog No. 602510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-251-0 10 mg
60-255-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-251-0 Supplier Tocris Bioscience™ Supplier No. 6025/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

MTH1 inhibitor

Potent MTH1 inhibitor (IC50 = 72 nM). Induces DNA damage and disrupts nucleotide pool homeostasis in cancer cells. Also attenuates colony formation of KRAS-mutated PANC1 cells in vitro. Suppresses tumor growth ∽50% in a colon cancer carcinoma xenograft model.

Chemical Identifiers

CAS 1374356-45-2
Molecular Formula C21H22Cl2FN5O

Specifications

Quantity 10 mg
Solubility Soluble to 50mM in DMSO and to 20mM in 1eq. HCl with gentle warming
Formula Weight 450.34
Purity >98%
Molecular Weight (g/mol) 450.339
Chemical Name or Material 3-[(1S)-1-(2,6-Dichloro-3-fluorophenyl)ethoxy]-5-[1-(4-piperidinyl)-1H-pyrazol-4-yl]-2-pyridinamine
Recommended Storage Store at RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.