Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S-Curve™ Safety Glass Dispenser
$179.20 - $233.50
Specifications
Color | Clear |
---|---|
Product Type | Safety Glasses Dispenser |
Dimensions (L x W x H) | 7 x 24.5 x 5 in. |
For Use With (Application) | Holds up to 7 pairs of Glasses |
Mounting | Wall Mount |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
19-162-410
|
S-Curve™
SGD07 |
Acrylic |
Each for $179.20
|
|
|||||
19-170-657
|
S-Curve™
SGD07PETG |
PETG |
Each for $233.50
|
|
|||||
Specifications
Clear | |
7 x 24.5 x 5 in. | |
Wall Mount |
Safety Glasses Dispenser | |
Holds up to 7 pairs of Glasses |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title