Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (S)-(+)-Dimethindene maleate
GREENER_CHOICE

Catalog No. 142510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-251-0 10 mg
14-255-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-251-0 Supplier Tocris Bioscience™ Supplier No. 1425/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

M2-selective antagonist

(S)-(+)-Dimethindene maleate is an enantiomer that is a subtype-selective M2 muscarinic receptor antagonist (pKi values are 7.08, 7.78, 6.70 and 7.00 for M1, M2, M3 and M4 receptors respectively). Also H1 histamine receptor antagonist (pKi = 7.48). Allows formation of extended pluripotent stem (EPS) cells in combination with CHIR 99021 (Cat.No. 4423), minocycline hydrochloride (Cat.No. 3268) and human leukemia inhibitory factor.

For more information about how Dimethindene maleate may be used, see our protocol: Generation and Propagation of EPS cells (LCDM Cocktail) .

Chemical Identifiers

CAS 136152-65-3
Molecular Formula C24H28N2O4

Specifications

Quantity 10 mg
Formula Weight 408.5
Purity >99%
Molecular Weight (g/mol) 408.498
Chemical Name or Material (S)-(+)-Dimethindene maleate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.