Learn More
Description
For more information about how Dimethindene maleate may be used, see our protocol: Generation and Propagation of EPS cells (LCDM Cocktail) .
Specifications
Specifications
| Quantity | 50 mg |
| Product Line | Tocris |
| Formula Weight | 408.5 |
| Purity | >99% |
| Molecular Weight (g/mol) | 408.498 |
| Chemical Name or Material | (S)-(+)-Dimethindene maleate |
Safety and Handling
| Recommended Storage | Desiccate at Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

