Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (S)-Duloxetine hydrochloride
GREENER_CHOICESDP

Catalog No. 479810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-981-0 10 mg
47-985-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-981-0 Supplier Tocris Bioscience™ Supplier No. 4798/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent 5-HT and NA reuptake inhibitor (SNRI); also blocks dopamine reuptake

High affinity, competitive 5-HT and noradrenaline (NA) re-uptake inhibitor (Ki values are 8.5 and 45 nM for 5-HT and NA reuptake respectively in cortical synaptosomes; IC50 values are 28 and 46 nM for 5-HT and NA reuptake respectively in rat hippocampal slices). Also blocks dopamine reuptake (Ki = 300 nM in striatal synaptosomes). Exhibits antidepressant and anxiolytic effects. Orally bioavailable.

Chemical Identifiers

CAS 136434-34-9
Molecular Formula C18H20ClNOS

Specifications

Quantity 10 mg
Formula Weight 333.88
Purity >99%
Molecular Weight (g/mol) 333.87
Chemical Name or Material (S)-Duloxetine hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.