Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Ethyl-N-[4-(trifluoromethyl)phenyl]-isothiourea hydrochloride, 98%, Thermo Scientific™

Catalog No. AC328910050
Click to view available options
Quantity:
5 mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163490-78-6
Molecular Formula C10H12ClF3N2S
Molecular Weight (g/mol) 284.725
MDL Number MFCD01320531
InChI Key UVJQIYZYQQKIAC-UHFFFAOYSA-N
Synonym tfpi hydrochloride, etpi hcl, s-ethyl-n-4-trifluoromethyl phenyl isothiourea hydrochloride, s-ethyl n-4-triflurormethyl phenyl isothio urea, hydrochloride, d01rgm, c10h11f3n2s.hcl, ethyl n'-4-trifluoromethyl phenyl carbamimidothioate hydrochloride, carbamimidothioic acid,n-4-trifluoromethyl phenyl-,ethyl ester,hydrochloride 1:1, carbamimidothioic acid,n-4-trifluoromethyl phenyl-, ethyl ester, hydrochloride 1:1
PubChem CID 2733516
IUPAC Name ethyl N'-[4-(trifluoromethyl)phenyl]carbamimidothioate;hydrochloride
SMILES CCSC(=NC1=CC=C(C=C1)C(F)(F)F)N.Cl

Specifications

Melting Point 143°C to 147°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 5 mg
Formula Weight 284.73
Percent Purity ≥97.5%
Physical Form Solid
Chemical Name or Material S-Ethyl-N-[4-(trifluoromethyl)phenyl]-isothiourea hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.