Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Ethyl thioacetate, 98+%

Catalog No. p-7080904
Click to view available options
Quantity:
5 g

CAS: 625-60-5 | C4H8OS | 104.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-60-5
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.167
MDL Number MFCD00015178
InChI Key APTGPWJUOYMUCE-UHFFFAOYSA-N
Synonym s-ethyl thioacetate, ethanethioic acid, ethyl ester, ethanethioic acid, s-ethyl ester, ethyl thiolacetate, ethyl ethanethioate, ethanethioic acid s-ethyl ester, s-ethyl thiolacetate, acetic acid, thio-, ethyl ester, thioethyl compound, acetic acid, thio-, s-ethyl ester
PubChem CID 61171
IUPAC Name S-ethyl ethanethioate
SMILES CCSC(=O)C

Specifications

Density 0.98
Boiling Point 116°C to 117°C
Flash Point 20°C (68°F)
Odor Stench
Refractive Index 1.46
Quantity 5 g
UN Number UN1993
Beilstein 1737643
Solubility Information Insoluble in water
Formula Weight 104.18
Percent Purity ≥98%
Chemical Name or Material S-Ethyl thioacetate
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 210-904-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.