Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Ethyl Trifluorothioacetate 99%, Thermo Scientific™

Catalog No. AC175880250
Click to view available options
Quantity:
5 mL
25 mL

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 383-64-2
Molecular Formula C4H5F3OS
Molecular Weight (g/mol) 158.138
InChI Key VGGUKFAVHPGNBF-UHFFFAOYSA-N
Synonym s-ethyl trifluorothioacetate, ethanethioic acid, trifluoro-, s-ethyl ester, ethyl trifluorothiolacetate, s-ethyl trifluoroethanethioate, trifluoroacetonylmercaptoethanol, acetic acid, trifluorothio-, s-ethyl ester, s-ethylthiotrifluoroacetate, trifluorothioacetic acid s-ethyl ester, ethanethioic acid, 2,2,2-trifluoro-, s-ethyl ester, 1-ethylsulfanyl-2,2,2-trifluoroethanone
PubChem CID 67844
IUPAC Name S-ethyl 2,2,2-trifluoroethanethioate
SMILES CCSC(=O)C(F)(F)F

Specifications

Color Colorless to Yellow
Density 1.23
Infrared Spectrum Authentic
Refractive Index 1.3762 to 1.3782 (20°C, 589nm)
Linear Formula CF3COSC2H5
Quantity 25 mL
Formula Weight 158.14
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material S-Ethyl trifluorothioacetate, 99%
EINECSNumber 206-852-1
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.