Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Furfuryl Thioformate, TCI America™
SDP

Catalog No. F068825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F068825G 25 g
F06885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F068825G Supplier TCI America Supplier No. F068825G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 59020-90-5
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.17
MDL Number MFCD00209507
InChI Key GFAOAYJTEVHTLA-UHFFFAOYSA-N
Synonym Thioformic Acid S-Furfuryl Ester, 2-(Formylthiomethyl)furan
PubChem CID 62144
IUPAC Name {[(furan-2-yl)methyl]sulfanyl}carbaldehyde
SMILES O=CSCC1=CC=CO1

Specifications

Color Yellow
Boiling Point 66°C
Quantity 25 g
Formula Weight 142.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material S-Furfuryl Thioformate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.