Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-Glycidyl Methyl Ether 97.0+%, TCI America™
SDP

Catalog No. G028125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G028125G 25 g
G02815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G028125G Supplier TCI America Supplier No. G028125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64491-68-5
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00273366
InChI Key LKMJVFRMDSNFRT-SCSAIBSYSA-N
Synonym (S)-1-Methoxy-2,3-epoxypropane, (S)-2-(Methoxymethyl)oxirane
PubChem CID 6992187
IUPAC Name (2S)-2-(methoxymethyl)oxirane
SMILES COCC1CO1

Specifications

Color Colorless
Quantity 25 g
UN Number 1992
Formula Weight 88.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-Glycidyl Methyl Ether
EINECSNumber (2)-2425
RTECSNumber TZ3530000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.