Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-Ibuprofen, 99%, Thermo Scientific Chemicals

Catalog No. p-147922
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

(S)-(+)-Ibuprofen, CAS # 51146-56-6, is the active enantomerof the non-steroidal anti-inflammatory drug, ibuprofen.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ibuprofen is a nonsteroidal anti-inflammatory drug commonly used in its racemic form
  • (S)-(+)-Ibuprofen is the active enantiomer of this drug that confers anti-inflammatory activity
  • (S)-(+)-Ibuprofen inhibits prostaglandin synthesis and thus reduces inflammation and pain when ingested

Applications

  • As the active enantiomer of ibuprofen, (S)-(+)-Ibuprofen is used in laboratory research for pharmacokinetic studies
  • (S)-(+)-Ibuprofen may be used as an inhibitor of cycloogenase 1 and 2 and an agonist of PPARalpha and PPARgamma
  • (S)-(+)-Ibuprofen may be used as material to study development of drug delivery systems
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51146-56-6
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.28
MDL Number MFCD00069289
InChI Key HEFNNWSXXWATRW-JTQLQIEISA-N
Synonym s-+-ibuprofen, dexibuprofen, s +-ibuprofen, s-2-4-isobutylphenyl propanoic acid, s-ibuprofen, d-ibuproten, s-+-2-4-isobutylphenyl propionic acid, s-+-4-isobutyl-alpha-methylphenylacetic acid, +-s-p-isobutylhydratropic acid, seractil
PubChem CID 39912
ChEBI CHEBI:43415
IUPAC Name (2S)-2-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)CC1=CC=C(C=C1)C(C)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 49.0°C to 53.0°C
Color White
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition + 57.00 (20.00°C c=2,EtOH)
Specific Rotation + 57.00
Solubility Information Solubility in water: insoluble. Other solubilities: readily soluble in most organic solvents
Formula Weight 206.28
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (S)-(+)-Ibuprofen, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.