Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Methyl-L-cysteine, 99+%, Thermo Scientific™

Catalog No. AC226920050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226920050 5 g Glass bottle
AC226920250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC226920050 Supplier Thermo Scientific Chemicals Supplier No. 226920050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1187-84-4
Molecular Formula C4H9NO2S
Molecular Weight (g/mol) 135.18
MDL Number MFCD00002612
InChI Key IDIDJDIHTAOVLG-UHFFFAOYNA-N
Synonym s-methyl-l-cysteine, s-methylcysteine, s-methyl-cysteine, h-cys me-oh, cysteine, s-methyl, l-cysteine, s-methyl, r-2-amino-3-methylthio propanoic acid, l-methylcysteine, unii-a34i1h07ym, ccris 1972
PubChem CID 24417
ChEBI CHEBI:45658
IUPAC Name 2-amino-3-(methylsulfanyl)propanoic acid
SMILES CSCC(N)C(O)=O

Specifications

Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3SCH2CH(NH2)CO2H
Quantity 5 g
Specific Rotation Condition − 29.50 (20.00°C c=1,water)
Specific Rotation -28.5° to -30.5° (20°C, 589nm) (c=1, H2O)
Formula Weight 135.18
Percent Purity ≥99.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material S-Methyl-L-cysteine
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.