Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Methyl thiobutyrate, 98%

Catalog No. p-7026239
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAB2214406

Supplier: Thermo Scientific Chemicals B2214406

Only null left
Add to Cart

CAS: 2432-51-1 | C5H10OS | 118.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2432-51-1
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD00009872
InChI Key GRLJIIJNZJVMGP-UHFFFAOYSA-N
Synonym s-methyl thiobutyrate, butanethioic acid, s-methyl ester, methyl thiolbutyrate, methanethiol butyrate, methanethiol n-butyrate, butyric acid, thio-, s-methyl ester, s-methyl thiobutanoate, unii-2p1e432myz, methyl thiobutanoate, methyl thiobutyrate natural
PubChem CID 62444
IUPAC Name S-methyl butanethioate
SMILES CCCC(=O)SC

Specifications

Density 0.966
Boiling Point 142°C to 143°C
Flash Point 34°C (93°F)
Refractive Index 1.462
Quantity 5 g
UN Number UN1993
Beilstein 1848987
Formula Weight 118.2
Percent Purity 98%
Chemical Name or Material S-Methyl thiobutyrate
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 219-407-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.