Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-tert-Butyldecahydroisoquinoline-3-carboxamide 97.0+%, TCI America™
SDP

Catalog No. B341025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B341025G 25 g
B34105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B341025G Supplier TCI America Supplier No. B341025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 136465-81-1
Molecular Formula C14H26N2O
Molecular Weight (g/mol) 238.375
MDL Number MFCD01313226
InChI Key UPZBXVBPICTBDP-UBNQGINQSA-N
Synonym s-n-tert-butyl decahydroisoquinoline-3-carboxamide
PubChem CID 123134381
IUPAC Name (4aR,8aR)-N-tert-butyl-1,2,3,4,4a,5,6,7,8,8a-decahydroisoquinoline-3-carboxamide
SMILES CC(C)(C)NC(=O)C1CC2CCCCC2CN1

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 25 g
Formula Weight 238.38
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-N-tert-Butyldecahydroisoquinoline-3-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.