Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S Opsin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
| Antigen | S Opsin |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
S Opsin Polyclonal specifically detects S Opsin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| S Opsin | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| 611 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RALKAVAAQQQESATTQKAEREVSRM | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Vision | |
| BCPBlue cone photoreceptor pigmentBlue-sensitive opsin, BOPshort-wave-sensitive opsin 1, CBT, opsin 1 (cone pigments), short-wave-sensitive | |
| OPN1SW | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title