Learn More
S Opsin Polyclonal Antibody, Rockland™

Goat Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | S Opsin |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1599
![]() ![]() |
Rockland Immunochemicals
600101MP7 |
100 μg |
Each for $636.24
|
|
|||||
50-199-1600
![]() ![]() |
Rockland Immunochemicals
600101MP7S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against mouse S Ospin. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence to mouse.
Opsin Blue belongs to the G-protein coupled receptor 1 family, opsin subfamily. It encodes the blue cone pigment gene which is one of three types of cone photoreceptors responsible for normal color vision. Defects in this gene are the cause of tritan color blindness (tritanopia). Affected individuals lack blue and yellow sensory mechanisms while retaining those for red and green. Defective blue vision is characteristic.Specifications
S Opsin | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Goat | |
Mouse | |
P51491 | |
12057 | |
Primary | |
Antibody |
1 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
AW551857; BCP; blue cone opsin; blue cone opsin-like pigment; Blue cone photoreceptor pigment; blue cone pigment; blue opsin; blue/UV opsin; Blue-sensitive opsin; BOP; CBT; OPN1SW; opsin 1 (cone pigments), short-wave-sensitive; opsin 1 (cone pigments), short-wave-sensitive (color blindness, tritan); S opsin; short wavelength sensitive opsin; Short wavelength-sensitive cone opsin; Short-wave-sensitive opsin 1; SWS opsin; UV cone pigment | |
OPN1SW | |
-20°C, Avoid Freeze/Thaw Cycles | |
OPN1SW |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.