Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S-Peptide Epitope Tag Antibody (6.2), Novus Biologicals™

Mouse Monoclonal Antibody
$173.10 - $487.50
Specifications
Antigen | S-Peptide Epitope Tag |
---|---|
Clone | 6.2 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
S-Peptide Epitope Tag Monoclonal antibody specifically detects S-Peptide Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.Specifications
S-Peptide Epitope Tag | |
1 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
IgG2b | |
Protein G purified |
6.2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Synthetic Peptide: K(1) E T A A A K F E R Q H M D S(15) C | |
Primary | |
This antibodyDetects the S-peptide in human samples. has been successfully used in Western blot, immunohistochemistry and immunofluorescence procedures. By Western blot this antibodyDetects the S-peptide in K562 cells. In immunohistochemistry, and immunofluorescence procedures recognizes the S-peptide in transfected K562 cells. The immunogen corresponds to the S-peptide conjugated through a C-terminal cysteine to keyhole limpet hemocyanin. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title