Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ S-peptide Epitope Tag Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581631 Supplier Invitrogen™ Supplier No. PA581631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Recognize N-terminal and C-terminal S tag in fusion proteins.

The S-Peptide epitope tag (S-Tag (TM)) can be used for numerous applications which include the analysis of localization, expression detection, and purification of polypeptides from mammalian cells. The small size of the tag (15 aa), the ability to purify tagged polypeptides without antibodies, and the very low expression of endogenous tissues in mammalian cells that react with S-Peptide make the S-Peptide antibody an attractive alternative to many existing epitope tags. Epitope tagging is a powerful and versatile strategy for detecting and purifying proteins expressed by cloned genes. To utilize this feature, protein expression vectors are typically engineered with a nucleotide sequence that encodes the peptide epitope tag. The gene of interest is cloned in-frame relative to the tag and, upon expression, the protein of interest is synthesized as a fusion protein with the peptide tag. Fusion protein detection and/or purification is mediated by highly specific antibodies to the engineered peptide, thus eliminating the need for antibodies to proteins from each newly cloned gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S-peptide Epitope Tag
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias S-tag
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the S-tag sequence (KETAAAKFERQHMDS).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.