Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Phenyl-L-cysteine, 97%

Catalog No. p-7036697
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 34317-61-8 | C9H11NO2S | 197.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34317-61-8
Molecular Formula C9H11NO2S
Molecular Weight (g/mol) 197.252
MDL Number MFCD01318758
InChI Key XYUBQWNJDIAEES-QMMMGPOBSA-N
Synonym s-phenyl-l-cysteine, beta-phenylcysteine, r-2-amino-3-phenylthio propanoic acid, s-phenylcysteine, h-cys phenyl-oh, l-cysteine, s-phenyl, 4-thia-l-homophenylalanine, 3-phenylthio-l-alanine, 2r-2-amino-3-phenylsulfanylpropanoic acid, 2r-2-amino-3-phenylthiopropanoic acid
PubChem CID 119462
IUPAC Name (2R)-2-amino-3-phenylsulfanylpropanoic acid
SMILES C1=CC=C(C=C1)SCC(C(=O)O)N

Specifications

Melting Point 200°C (decomposition)
Beilstein 2268203
Quantity 1 g
Formula Weight 197.25
Optical Rotation +10° (c=1.5 in 1M NaOH)
Percent Purity 97%
Chemical Name or Material S-Phenyl-L-cysteine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.