Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-(+)-Prolinol, 98%

Click to view available options
Quantity:
1g
5g
25g
23356-96-9
C5H11NO
101.149
MFCD00005255
HVVNJUAVDAZWCB-YFKPBYRVSA-N
l-prolinol,l-+-prolinol,s-pyrrolidin-2-ylmethanol,s-+-2-pyrrolidinemethanol,s-prolinol,2s-pyrrolidin-2-ylmethanol,l +-prolinol,2s-pyrrolidin-2-yl methanol,s-+-2-hydroxymethyl pyrrolidine,s-+-prolinol
640091
[(2S)-pyrrolidin-2-yl]methanol
C1CC(NC1)CO
23356-96-9
C5H11NO
101.149
MFCD00005255
HVVNJUAVDAZWCB-YFKPBYRVSA-N
l-prolinol,l-+-prolinol,s-pyrrolidin-2-ylmethanol,s-+-2-pyrrolidinemethanol,s-prolinol,2s-pyrrolidin-2-ylmethanol,l +-prolinol,2s-pyrrolidin-2-yl methanol,s-+-2-hydroxymethyl pyrrolidine,s-+-prolinol
640091
[(2S)-pyrrolidin-2-yl]methanol
C1CC(NC1)CO
Applications
It is an active pharmaceutical intermediate.

Solubility
Fully miscible in water. Soluble in chloroform.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Melting Point 42°C to 44°C
Boiling Point 74°C to 76°C (2mmHg)
Quantity 1g
Flash Point 86°C (186°F)
Optical Rotation +31° (c=1 in Toluene)
Beilstein 79843
Formula Weight 101.15
Percent Purity 98%
Chemical Name or Material (S)-(+)-Prolinol

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning