Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S,S)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane 97.0+%, TCI America™
SDP

Catalog No. B3036100MG
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 97858-62-3
Molecular Formula C28H28O2P2
Molecular Weight (g/mol) 458.48
MDL Number MFCD05863547
InChI Key QKZWXPLBVCKXNQ-UHFFFAOYNA-N
Synonym 1,2-bis s-2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp, unii-hs6f5ew3u6, hs6f5ew3u6, ethylenebis s-o-anisyl phenyl phosphine, s,s-+-1,2-bis 2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp; s,s-ethylenebis 2-methoxyphenyl phenylphosphine, s-2-methoxyphenyl 2-s-2-methoxyphenyl phenyl phosphanyl ethyl phenylphosphane
PubChem CID 11419748
IUPAC Name (2-methoxyphenyl)({2-[(2-methoxyphenyl)(phenyl)phosphanyl]ethyl})phenylphosphane
SMILES COC1=CC=CC=C1P(CCP(C1=CC=CC=C1)C1=CC=CC=C1OC)C1=CC=CC=C1

Specifications

Melting Point 104°C
Quantity 100 mg
Formula Weight 458.48
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S,S)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.