Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S,S)-(-)-1,2-Diphenyl-1,2-ethanediol, +98%, Thermo Scientific™

Catalog No. AC279515000
Click to view available options
:
500mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2325-10-2
CAS Min % 98.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C14H14O2
Quantity 500mg
MDL Number MFCD00064255
Specific Rotation Condition − 93.00 (20.00°C c=2.5,ethanol)
Specific Rotation -92° to -94° (20°C, 589nm) (c=2.5, EtOH)
Synonym s,s---hydrobenzoin, 1s,2s-1,2-diphenylethane-1,2-diol, s,s-hydrobenzoin, s,s---1,2-diphenyl-1,2-ethanediol, hydrobenzoin,-, unii-wx45q7714b, 1,2-ethanediol, 1,2-diphenyl-, 1s,2s, s,s-1,2-diphenylethylene glycol, 1s,2s---1,2-diphenyl-1,2-ethanediol, isohydrobenzoin
InChI Key IHPDTPWNFBQHEB-KBPBESRZSA-N
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O
IUPAC Name (1S,2S)-1,2-diphenylethane-1,2-diol
Molecular Weight (g/mol) 214.26
PubChem CID 853020
Formula Weight 214.26
Percent Purity ≥98.0%
Chemical Name or Material (S,S)-(-)-1,2-Diphenyl-1,2-ethanediol, 98+%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.