Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S,S)-TaDiAS-2nd 95.0+%, TCI America™
SDP

Catalog No. D34761G
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 2135524-59-1
Molecular Formula C58H70B2F8N2O2
Synonym (S,S)-TaDiAS-2nd

Specifications

Color White-Yellow
Quantity 1 g
UN Number 1759
Formula Weight 1000.82
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 6,10-Dibenzyl-N,N′-dimethyl-N,N,N′,N′-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.