Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S,S)-Ts-DENEB(regR), TCI America™
SDP

Catalog No. p-7137562
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 1384974-37-1
Molecular Formula C31H33ClN2O3RuS
Molecular Weight (g/mol) 650.196
MDL Number MFCD22666040
InChI Key INKUCOHLIHBSDN-YBZGWEFGSA-M
Synonym Chloro[(S,S)-N-[2-[2-(4-methylbenzyloxy)ethyl]amino-1,2-diphenylethyl]-p-toluenesulfonamide]ruthenium(II), Chloro[(S,S)-N-[2-(4-methylbenzyloxy)ethyl]-N′C-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
PubChem CID 121235453
IUPAC Name chlororuthenium(1+);[(1S,2S)-2-[2-[(4-methylphenyl)methoxy]ethylamino]-1,2-diphenylethyl]-(4-methylphenyl)sulfonylazanide
SMILES CC1=CC=C(C=C1)COCCNC(C2=CC=CC=C2)C(C3=CC=CC=C3)[N-]S(=O)(=O)C4=CC=C(C=C4)C.Cl[Ru+]

Specifications

Quantity 200 mg
UN Number 3077
Formula Weight 650.20
Physical Form Crystalline Powder
Chemical Name or Material (S,S)-Ts-DENEB(regR)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.