Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ (S)-SNAP 5114
GREENER_CHOICESDP

Catalog No. 156150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-615-0 50 mg
15-611-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-615-0 Supplier R&D Systems™ Supplier No. 1561/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. GABA uptake inhibitor

GABA transport inhibitor, showing selectivity for GAT-3 and GAT-2 (IC50 values are 5, 21 and 388 μM for hGAT-3, rGAT-2 and hGAT-1 respectively). Increases thalamic GABA levels and is an anticonvulsant following systemic administration in vivo.

Chemical Identifiers

CAS 157604-55-2
Molecular Formula C30H35NO6

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 505.61
Purity >98%
Molecular Weight (g/mol) 505.61
Chemical Name or Material (S)-SNAP 5114
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.