Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ S-TEX 377SC Fully Foam Nitrile Coated Cut Resistant Gloves

Catalog No. 19021055 Shop All Showa Best Glove Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-021-055 X-Large
19-021-053 Medium
19-021-054 Large
19-021-056 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-021-055 Supplier SHOWA™ Supplier No. STEX377SCXL09
Add to Cart
Add to Cart
This item is not returnable. View return policy

Engineered with steel-reinforced Hagane Coil™ technology to provide ANSI A4 cut protection. Foam nitrile coating enhances grip capabilities, in addition to resisting hydrocarbons, grease, and oil.

  • Engineered with steel-reinforced Hagane Coil™ technology to provide ANSI A4 cut resistance
  • Lined with Kevlar™ aramid fibers for additional strength
  • Gauntlet style reinforced safety cuff extends cut protection up to the forearm
  • Nitrile coating is resistant to hydrocarbons, grease, and oil
  • Additional foam nitrile palm dip enhances wet and dry grip capabilities
  • Ergonomically designed hand mold reduces hand fatigue

Specifications

ANSI Abrasion Rating ANSI 6
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Cuff Material Reinforced
Coating Material Nitrile Foam
Color Black, Blue
Cut Resistance Rating ANSI A4
Exterior Finish Partially Textured
Hand Specific
Material Nylon, Polyester, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size X-Large
EN388 Standard 4X41D
For Use With (Application) Drainage, HVAC, Mechanical Work, and Window Manufacturing
Glove Cuff Style Safety
Liner Material Aramid Fiber
Puncture Rating ANSI/ISEA 105-16 Level 3
Series S-TEX 377SC
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.