Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

s-Trioxane, 99+%

Catalog No. p-3737693
Click to view available options
Quantity:
5 g
100 g
500 g
2 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 110-88-3 | C3H6O3 | 90.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-88-3
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00006563
InChI Key BGJSXRVXTHVRSN-UHFFFAOYSA-N
Synonym s-trioxane, trioxymethylene, metaformaldehyde, triformol, formaldehyde, trimer, aldeform, formagene, marvosan, trioxan, sym-trioxane
PubChem CID 8081
ChEBI CHEBI:38043
IUPAC Name 1,3,5-trioxane
SMILES C1OCOCO1

Specifications

Melting Point 61.0°C to 62.0°C
Color Colorless to White
Density 1.1700g/mL
Boiling Point 114.0°C to 116.0°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 19,381
Merck Index 15,9906
Specific Gravity 1.17
Solubility Information Solubility in water: soluble in water: 221g/L (25°C). Other solubilities: soluble in alcohols,ether,ketones,acetone,,chlorinatid and aromatic hydrocarbons,and,other organic solvents,slightly soluble in pentane and petr.ether
Sublimation Point 0115.0°C
Formula Weight 90.08
Percent Purity 99+%
Physical Form Crystalline Flakes or Crystals
Chemical Name or Material s-Trioxane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Suspected of damaging the unborn child.
Flammable solid.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-812-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.