Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100-A9 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984909
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4909 Supplier Bioss Supplier No. BS2697R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

S100A9 (MRP-14) is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis. Reconstitute with 1.0mL distilled water.

Specifications

Antigen S100-A9
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene S100A9
Gene Accession No. P06702
Gene Alias 60B8Ag; AI323541; AW546964; B8Ag; BEE22; BOS_2815; CAGB; calgranulin B; Calgranulin-B; calprotectin; Calprotectin L1H subunit; CFAG; CGLB; GAGB; intracellular calcium-binding protein (MRP14); L1AG; Leukocyte L1 complex heavy chain; LIAG; MAC387; MIF; Migration inhibitory factor-related protein 14; MRP14; MRP-14; Myeloid-related protein 14; Neutrophil cytosolic 23 kDa protein; NIF; P14; p23; Protein S100-A9; RNA-binding region containing protein 2-like; RNA-binding region containing protein 2-like; S100 calcium binding protein A9; S100 calcium binding protein A9; S100 calcium binding protein A9 (calgranulin B); S100 calcium-binding protein A9; S100 calcium-binding protein A9 (calgranulin B); S100a9
Gene Symbols S100A9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human S100-A9 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6280
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.