Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100 beta (S100B) Monoclonal Antibody (OTI2H10), TrueMAB™, OriGene
SDP

Catalog No. 502544098
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4098 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4098 Supplier OriGene Supplier No. CF800599
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100 beta (S100B) Monoclonal antibody specifically detects S100 beta (S100B) in Human samples. It is validated for ELISA

Specifications

Antigen S100 beta (S100B)
Applications ELISA
Classification Monoclonal
Clone OTI2H10
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene JUN
Gene Accession No. P05412
Gene Alias AI850290; Bpb; hypothetical protein LOC525716; NEF; protein S100-B; S100; S100 calcium binding protein B; S100 calcium binding protein beta (neural); S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S-100 calcium-binding protein beta subunit; S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S100 protein beta chain; S-100 protein beta chain; S-100 protein subunit beta; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100B; S100-B; S100beta; S100P
Host Species Mouse
Immunogen Full length human recombinant protein of human JUN produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3725
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.