Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100 calcium binding protein A14 Antibody (S100A14/7402), Novus Biologicals™

Mouse Monoclonal Antibody
$216.50 - $443.50
Specifications
Antigen | S100 calcium binding protein A14 |
---|---|
Clone | S100A14/7402 |
Concentration | 0.2 mg/ml |
Dilution | ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL |
Applications | ELISA, Immunohistochemistry (Paraffin) |
Description
S100 calcium binding protein A14 Monoclonal antibody specifically detects S100 calcium binding protein A14 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
S100 calcium binding protein A14 | |
0.2 mg/ml | |
ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cancer | |
10mM PBS with 0.05% BSA | |
57402 | |
IgG2b κ | |
Protein A or G purified |
S100A14/7402 | |
ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
protein S100-A14, S100 calcium binding protein A14, S100 calcium-binding protein A14, S100A15BCMP84breast cancer membrane protein 84, S114 | |
Recombinant full-length human S100 calcium binding protein A14 protein | |
Primary | |
Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title