Learn More
S100 Monoclonal Antibody (15E2E2), Abbomax
Mouse Monoclonal Antibody
Supplier: Abbomax Inc 5006824

Description
Positive control: Melanoma or Schwannoma Cellular location: Cytoplasmic.
S100 Monoclonal antibody specifically detects S100 in Bovine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western BlotSpecifications
S100 | |
Monoclonal | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
P02638, P02639, P04271, P04631, P23297, P35467, P50114, P56565 | |
Mouse | |
Protein A | |
RUO | |
20193, 20203, 25742, 295214, 525716, 528735, 6271, 6285 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot | |
15E2E2 | |
Unconjugated | |
S100A1 | |
AI266795; AI850290; Bpb; hypothetical protein LOC525716; NEF; Protein S100-A1; protein S100-B; S 100; S100; S100 alpha; S100 calcium binding protein A1; S100 calcium binding protein B; S100 calcium-binding protein A1; S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S-100 protein alpha chain; S-100 protein beta chain; S-100 protein subunit alpha; S-100 protein subunit beta; S100 protein, alpha polypeptide; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100a; S100A1; S100alpha; S100-alpha; S100B; S100-B; S100beta; S100P | |
Purified bovine brain S100 protein | |
100 μg | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.