Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100 Polyclonal Antibody, Abbomax

Catalog No. 502203644
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3644 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3644 Supplier Abbomax Inc Supplier No. 5002144
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Melanoma or Schwannoma Cellular location: Cytoplasmic.

S100 Polyclonal antibody specifically detects S100 in Bovine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot

Specifications

Antigen S100
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene S100A1
Gene Accession No. P02638, P02639, P04271, P04631, P23297, P35467, P50114, P56565
Gene Alias AI266795; AI850290; Bpb; hypothetical protein LOC525716; NEF; Protein S100-A1; protein S100-B; S 100; S100; S100 alpha; S100 calcium binding protein A1; S100 calcium binding protein B; S100 calcium-binding protein A1; S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S-100 protein alpha chain; S-100 protein beta chain; S-100 protein subunit alpha; S-100 protein subunit beta; S100 protein, alpha polypeptide; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100a; S100A1; S100alpha; S100-alpha; S100B; S100-B; S100beta; S100P
Host Species Rabbit
Immunogen Recombinant protein encoding human S100 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20193, 20203, 25742, 295214, 525716, 528735, 6271, 6285
Target Species Bovine, Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.